Order Entry
Spain
Orders LinkContactUsLinkComponent
Fluoro Risperidone, Pharmaceutical Analytical Impurities

Fluoro Risperidone, Pharmaceutical Analytical Impurities

5-Fluoro Risperidone, Pharmaceutical Analytical Impurities, Vial, 25 mg
Catalog # 1A00520.
Supplier:   USP
Fluoro Risperidone, Pharmaceutical Analytical Impurities
Catalog # 1A00520.
Supplier:  USP
CAS Number:  
Fluoro Risperidone, Pharmaceutical Analytical Impurities

5-Fluoro Risperidone, Pharmaceutical Analytical Impurities, Vial, 25 mg

CAS Number:  
Price
682,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Caution: Cold Shipment required

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • Description:
    5-Fluoro Risperidone
  • Cat. No.:
    1A00520.
  • Description:
    5-Fluoro Risperidone
  • Synonym(s):
    3-(2-(4-(5-fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
  • CAS No.:
    1199589-74-6

Specifications