DMP (Dimethyl pimelimidate dihydrochloride), Pierce™
:58537-94-3
:
:C₉H₁₈N₂O₂
:MFCD00038942
:259.17 g/mol
:Refrigerator
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Homobifunctional
- Reactive Moiety:Imidoester
- Spacer:Short
- Form:Powder
- Quantity:50 mg
- Type:Crosslinker
- Solubility:Water
- Spacer Arm:9.2 Å
Frequently Bought Together