Stainless steel lids for water baths
Supplier: NICKEL ELECTRO
Certificates
About this item
Stainless steel gable lids helps reduce evaporation, assists in keeping samples in a water bath free from contamination and saves energy.
- High profile allows extended use of the bath, right into the edges of the tank
- No drip points to contaminate samples whilst the lid design ensures all condensate returns back into the bath
- Insulated handles for safe use
Frequently Bought Together