Order Entry
Spain
ContactUsLinkComponent
SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene), Pierce™
SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene), Pierce™
Catalog # PIER21558
CAS Number:  
undefined
SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene), Pierce™
Catalog # PIER21558
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :360.49 g/mol
Storage Temperature:Refrigerator
CAS Number:112241-19-7
Formula:C₁₈H₂₀N₂O₂S₂
MDL Number:MFCD00083161