Anti-IRE1 alpha (phospho-Ser724) Rabbit Polyclonal Antibody
NOVUNB100-2323
- Antibody type:Primary
- Antigen name:IRE1 alpha
- Antigen symbol:IRE1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Primates,Pig,Rabbit,Monkey,Mammals,Human,Rat,Mouse
- Western blot:Yes
- Gene ID:2081
- Antigen synonyms:phospho-ire1 alpha|IRE 1a|ire1-alpha|Endoplasmic reticulum-to-nucleus signaling 1|ER to nucleus signalling 1|ire1 alpha phosphorylation|Inositol-requiring protein 1|inositol-requiring 1|hIRE1p|protein kinase/endoribonuclease|endoplasmic reticulum to nucleus signaling 1|IRE1P|ire1 alpha p Ser724|IRE1MGC163279|EC 2.7.11|p-ire1 alpha|serine/threonine-protein kinase/endoribonuclease IRE1|IRE1a|MGC163277|phospho ire1 alpha|phospho-ire1a|FLJ30999
- Modification:phospho-Ser724
- Storage buffer:PBS
- Storage temperature:Aliquot and store at −20 °C or -80C. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:A synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460]
- Purification:Immunogen affinity purified
- Pk:0,1 mL
Rabbit Polyclonal IRE1 alpha [p Ser724] Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate (-).
Type: Primary
Antigen: IRE1 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human