Order Entry
Spain
ContactUsLinkComponent
Anti-Ajuba Rabbit Polyclonal Antibody
Anti-Ajuba Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-89570
Supplier:  Novus Biologicals
Anti-Ajuba Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-89570
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Ajuba
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    84962
  • Antigen synonyms:
    jub|ajuba homolog (Xenopus laevis)|protein ajuba|Ajuba|MGC15563
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:LDRLYAQRPAGFGCQESRHSYPPALGSPGALAGAGVGAAGPLERRGAQPGRHSVTGYGDCAVGARYQDELTALLRLTVGTGGREAGARGEPSGIEPSGLEEPPGPFVPEAARARMREPEAREDYFGTCIKCNKGIYGQSN
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μl
  • Pk:
    0,1 mL

Specifications

About this item

The Ajuba Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ajuba. This antibody reacts with human. The Ajuba Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Ajuba
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human