Order Entry
Spain
ContactUsLinkComponent
MEM Nonessential amino acids, Corning®
MEM Nonessential amino acids, Corning®
Catalog # 25-025-CI
Supplier:  Corning
CAS Number:  
undefined
MEM Nonessential amino acids, Corning®
Catalog # 25-025-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Source:
  • Species:
  • Description:
    MEM Nonessential amino acids, 1 L
  • Seq:
    12
  • Environmentally Preferable:
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen

Specifications

About this item

The addition of MEM nonessential amino acids to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells.

This concentrated supplement allows for the efficient addition to media with no risk of dilution of media components.