Dimethyl sulphoxide, sterile filtered
Catalog # 25-950-CQC
Supplier: Corning
CAS Number:
Specifications
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:78.14 g/mol
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089