Order Entry
Spain
ContactUsLinkComponent
Bacillus cereus WDCM 00001 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Bacillus cereus WDCM 00001 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000013-10EA
Supplier:  Merck
CAS Number:  
Bacillus cereus WDCM 00001 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000013-10EA
Supplier:  Merck
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pk:
    10
  • Description:
    Bacillus cereus WDCM 00001 Vitroids™, 80-130 CFU mean value range
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    −20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

These Vitroids™ discs contain Bacillus cereus WDCM 00001 viable microorganisms (15-80 CFU) in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.

  • Defined CFU ranges and low standard deviation
  • Fast, reliable and easy to use

The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.