Order Entry
Spain
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), powder, Corning®
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), powder, Corning®
Catalog # MDTC46-101-RF
Supplier:  Corning
CAS Number:  
undefined
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), powder, Corning®
Catalog # MDTC46-101-RF
Supplier:  Corning
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :408.63 g/mol
Melting Pt:237…239 °C
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666