Anti-CD184 (CXCR4) Mouse Monoclonal Antibody [clone: 12G5] (APC (Allophycocyanin)/FIRE™750)
Catalog # BLEG306538
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:CD184 (CXCR4)
- Clonality:Monoclonal
- Clone:12G5
- Conjugation:APC (Allophycocyanin)/FIRE™750
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Baboon,African green monkey,Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Chimpanzee,Human
- Antigen synonyms:CXCR4|Fusin
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:2…8 °C
- Immunogen:CP-MAC-infected Sup-T1 cells
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with APC/Fire™ 750 under optimal conditions.
- Size:100 tests
- Pk:100 Tests