CF® Dye BCN
About this item
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labelling reactions are also available.
- Fluorescent labelling of azide groups on target molecules via copper-free bioorthonal chemistry
- Alternative to fluorescently labelled DIBO and DBCO
- Suitable for surface or intracellular labelling of live cells or cell extracts
- Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labelling
- Available in several CF® Dye options including membrane-permeant forms for intracellular labelling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labelled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labelling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labelling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium's next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
12 Options Available Below
Product Details & Documents
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labelling reactions are also available.
- Fluorescent labelling of azide groups on target molecules via copper-free bioorthonal chemistry
- Alternative to fluorescently labelled DIBO and DBCO
- Suitable for surface or intracellular labelling of live cells or cell extracts
- Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labelling
- Available in several CF® Dye options including membrane-permeant forms for intracellular labelling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labelled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labelling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labelling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium's next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Product Family Options
Product Information
- Ex/EmPkAvailabilityPrice
- 92114.CF®405M BCN, membrane permeability: ImpermeantCF®405M BCN, membrane permeability: Impermeant408/452 nm0,5 mgLog in for availability1 * 0,5 mg611,00 €
- 92113.CF®405S BCN, membrane permeability: ImpermeantCF®405S BCN, membrane permeability: Impermeant404/431 nm0,5 mgLog in for availability1 * 0,5 mg611,00 €
- 92075.CF®488A BCN, membrane permeability: ImpermeantCF®488A BCN, membrane permeability: Impermeant490/515 nm0,5 mgLog in for availability1 * 0,5 mg601,00 €
- 92076.CF®568 BCN , membrane permeability: ImpermeantCF®568 BCN , membrane permeability: Impermeant562/583 nm0,5 mgLog in for availability1 * 0,5 mg601,00 €
- 92077.CF®594 BCN, membrane permeability: ImpermeantCF®594 BCN, membrane permeability: Impermeant593/614 nm0,5 mgLog in for availability1 * 0,5 mg601,00 €
- 92078.CF®640R BCN, membrane permeability: ImpermeantCF®640R BCN, membrane permeability: Impermeant642/662 nm0,5 mgLog in for availability1 * 0,5 mg601,00 €
- 96059.CF®647 BCN, membrane permeability: ImpermeantCF®647 BCN, membrane permeability: Impermeant650/665 nm0,5 mgLog in for availability1 * 0,5 mg575,00 €
- 96058.CF®680 BCN , membrane permeability: ImpermeantCF®680 BCN , membrane permeability: Impermeant681/698 nm0,5 mgLog in for availability1 * 0,5 mg575,00 €
- 92079.CF®680R BCN , membrane permeability: ImpermeantCF®680R BCN , membrane permeability: Impermeant680/701 nm0,5 mgLog in for availability1 * 0,5 mg601,00 €
- 96070.CF®440 BCN , membrane permeability: PermeantCF®440 BCN , membrane permeability: Permeant440/515 nm0,5 mgLog in for availability1 * 0,5 mg575,00 €
- 96026.CF®500 BCN, membrane permeability: PermeantCF®500 BCN, membrane permeability: Permeant502/511 nm0,5 mgLog in for availability1 * 0,5 mg592,00 €
- 96027.CF®650 BCN, membrane permeability: PermeantCF®650 BCN, membrane permeability: Permeant653/673 nm0,5 mgLog in for availability1 * 0,5 mg592,00 €
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.