Order Entry
Spain
ContactUsLinkComponent
Absolutely total RNA purification kits
Absolutely total RNA purification kits
Supplier:  AGILENT
Certificates
Absolutely total RNA purification kits
Supplier:  AGILENT

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Sample type:
    Human/animal cells and tissues
  • Target molecule:
    RNA

Specifications

Frequently Bought Together