Sample introduction supplies for perkinElmer
HEWL8003-0414
: Agilent
- Description:Y-connector
- Environmentally Preferable:
- Pk:1
These sample introduction supplies are equivalent to PerkinElmer AA supplies, so you can be confident that they will integrate seamlessly with PerkinElmer AA system.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure to get the best performance from PerkinElmer system when using parts and supplies.