GC columns, J&W DB-Dioxin
Supplier: Agilent
Certificates
About this item
DB-Dioxin is specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDD) and dibenzofurans (PCDF).
- Specifically engineered for the analysis of polychlorinated dibenzodioxins (PCDDs) and dibenzofurans (PCDFs)
- Resolves 2,3,7,8-TCDD and 2,3,7,8-TCDF from all other isomers in one run
- Low bleed
- Bonded and cross-linked
- Solvent rinsable
A 100-psi regulator is required to reach optimum carrier gas velocity.
Frequently Bought Together