J.T.Baker®, Plate 384
Supplier: Avantor
Certificates
About this item
Available in PP for automated screening and storage systems.
- Alphanumeric code to assist sample identification
- Square wells for increased volume
- V-bottom for high sample recovery
- High frame (3 mm) for stable and slip-free stacking
J.T.Baker Plate 384 is ideal for small sample volumes.
Specifications
- Catalog No:
- 43004-0200
- 43004-0120
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)