Order Entry
Spain
ContactUsLinkComponent
STERI-OIL® 200
STERI-OIL® 200
Catalog # VELTSO200-A1Z
Supplier:  VELTEK ASSOCIATES
undefined
STERI-OIL® 200
Catalog # VELTSO200-A1Z
Supplier:  VELTEK ASSOCIATES
Specifications
  • Description:
    Sterile mineral oil, STERI-OIL 200, dropper bottle, size: 1 oz.
  • Pk:
    250
Specifications
About this item
STERI-OIL 200 is a sterile mineral oil lubricant used for lubricating machinery in aseptic areas. The heavy consistency provides lubrication and prevents metal to metal contact, withstands high friction without displacement, and reduces metal fatigue.