Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Spain
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Hepatitis E Virus ORF2 (452-617)
Recombinant Hepatitis E Virus ORF2 (452-617)
Recombinant Hepatitis E Virus ORF2 (452-617)
APOSBITP1830-1MG
:
Apollo Scientific
:
Recombinant Hepatitis E Virus ORF2 (452-617)
APOSBITP1830-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HEV ORF2
Protein/peptide name:
Hepatitis E Virus ORF2 (452-617)