Order Entry
Spain
ContactUsLinkComponent
Recombinant Hepatitis E Virus ORF2  (633-659)
Recombinant Hepatitis E Virus ORF2  (633-659)
Catalog # APOSBITP1828-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis E Virus ORF2  (633-659)
Catalog # APOSBITP1828-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HEV ORF2
  • Protein/peptide name:
    Hepatitis E Virus ORF2  (633-659)

Specifications

About this item