0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Spain
ContactUsLinkComponent
Proteins and Peptides
Rat Recombinant Leptin tA
Rat Recombinant Leptin tA
APOSBITP1208-1MG
:
Apollo Scientific
:
Rat Recombinant Leptin tA
APOSBITP1208-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Rat
Protein synonyms:
Leptin Triple Antagonist
Protein/peptide name:
Leptin tA