Bovine recombinant FGF-b
Catalog # APOSBITP1049-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Bovine
- Protein synonyms:Bovine Fibroblast Growth Factor-basic
- Protein/peptide name:FGF-b