Human recombinant SDF-1b
Catalog # APOSBITC2046-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Seq:23
- Protein synonyms:Stromal Cell-Derived Factor-1 beta, CXCL12
- Protein/peptide name:SDF-1b