Order Entry
Spain
ContactUsLinkComponent
Rat recombinant MIP-1a
Rat recombinant MIP-1a
Catalog # APOSBITC2036-1MG
Supplier:  Apollo Scientific
CAS Number:  
Rat recombinant MIP-1a
Catalog # APOSBITC2036-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Protein synonyms:
    Macrophage Inflammatory protein-1 alpha, CCL3
  • Protein/peptide name:
    MIP-1a

Specifications

About this item