Human recombinant MCP-1
Catalog # APOSBITC2024-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Monocyte Chemotactic Protein-1/MCAF, CCL2
- Protein/peptide name:MCP-1