
Difluoroacetic acid ≥97.5% (by GC), LiChropur™ for LC-MS, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:381-73-7
:
Formula:C₂H₂F₂O₂
MDL Number:MFCD00004220
:96.03 g/mol
Boiling Pt:134 °C (1013 hPa)
Melting Pt:–1 °C
Density:1.526 g/cm³ (20 °C)
EINECS:206-839-0
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:3265
:GHS05
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together