Order Entry
Spain
ContactUsLinkComponent
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene 97%
Irritant
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene 97%
Catalog # APOSPC9642-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Trifluoro(1,1,2,2-tetrafluoro-2-bromoethoxy)ethylene 97%
Catalog # APOSPC9642-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :276.94 g/mol
Boiling Pt:55 °C
EINECS:288-516-4
 :GHS07
Storage Temperature:Refrigerator
CAS Number:85737-06-0
Formula:F₂CCFOCF₂CF₂Br
MDL Number:MFCD02183517