

3-Fluoro-4-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:247113-90-2
:
Formula:C₉H₅F₄N
MDL Number:MFCD00236305
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together