

HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:148893-10-1
:
Formula:C₁₀H₁₅F₆N₆OP
MDL Number:MFCD00274639
:380.2 g/mol
Melting Pt:186 °C (dec.)
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together