2,2,2-Trifluoroethanol 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75-89-8
:
Formula:C₂H₃F₃O
MDL Number:MFCD00004672
:100.04 g/mol
Boiling Pt:74…75 °C
Melting Pt:–44 °C
Density:1.373 g/cm³ (25 °C)
Flash Pt:29 °C
EINECS:200-913-6
UN:1986
ADR:3+6.1,III
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Frequently Bought Together