Order Entry
Spain
ContactUsLinkComponent
3,5-Bis(trifluoromethyl)benzyl chloride 98%
Corrosive
3,5-Bis(trifluoromethyl)benzyl chloride 98%
Catalog # APOSPC3105G-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3,5-Bis(trifluoromethyl)benzyl chloride 98%
Catalog # APOSPC3105G-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.58 g/mol
Boiling Pt:46 °C/3mmHg
Melting Pt:29…33 °C
Flash Pt:71 °C
Storage Temperature:Ambient
EINECS:278-216-1
UN:3261
ADR:8,II
 :GHS05
CAS Number:75462-59-8
Formula:C₉H₅ClF₆
MDL Number:MFCD00009906