
2'-Chloro-5'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71648-45-8
:
Formula:C₉H₆ClF₃O
MDL Number:MFCD00045092
:222.59 g/mol
Boiling Pt:98…100 °C/6mmHg
Density:1.392 g/cm³ (25 °C)
Flash Pt:91.1 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together