
6-Chloro-2-(trifluoromethyl)nicotinic acid
Catalog # APOSPC201216-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo de env.:Bottle
- Env:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:160…162 °C
:225.56 g/mol
:GHS07
Storage Temperature:Ambiente
CAS Number:261635-83-0
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD11840271
Storage Temperature:Ambient