
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:128625-52-5
:
Formula:C₁₈H₂₈F₆N₆OP₂
MDL Number:MFCD00077411
:520.4 g/mol
Melting Pt:154…156 °C (dec.)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together