3-Fluoro-4-(trifluoromethoxy)phenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:187804-79-1
:
Formula:C₇H₅BF₄O₃
MDL Number:MFCD07368680
:223.920 g/mol
Melting Pt:144…149 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together