
N-(tert-Butoxycarbonyl)-D-threonine methyl ester 98+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96099-84-2
:
Formula:C₁₀H₁₉NO₅
MDL Number:MFCD01073763
:233.26 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together