Order Entry
Spain
ContactUsLinkComponent
3,6-Bis(hydroxymethyl)durene 95%
3,6-Bis(hydroxymethyl)durene 95%
Catalog # APOSOR936985-5G
Supplier:  Apollo Scientific
CAS Number:  
3,6-Bis(hydroxymethyl)durene 95%
Catalog # APOSOR936985-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.27 g/mol
Storage Temperature:Ambient
CAS Number:7522-62-5
Formula:C₁₂H₁₈O₂
MDL Number:MFCD00004623