Order Entry
Spain
ContactUsLinkComponent
Dimethyl succinate 95%
Dimethyl succinate 95%
Catalog # APOSOR923432-100G
Supplier:  Apollo Scientific
CAS Number:  
Dimethyl succinate 95%
Catalog # APOSOR923432-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :146.14 g/mol
Storage Temperature:Ambient
CAS Number:106-65-0
Formula:C₆H₁₀O₄
MDL Number:MFCD00008466