Dichloroacetic acid (DCA) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:79-43-6
:
Formula:C₂H₂Cl₂O₂
MDL Number:MFCD00004223
:128.94 g/mol
Storage Temperature:Ambient
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Bromine ≥99.8%, liquid
Supplier: Thermo Fisher Scientific
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% unstabilised, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99.5% for analysis
Supplier: Thermo Fisher Scientific