Aniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:142-04-1
:
Formula:C₆H₈ClN
MDL Number:MFCD00012958
Boiling Pt:245 °C
Melting Pt:196…200 °C
Density:1.221 g/cm³ (25 °C)
Flash Pt:193 °C
UN:1548
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
:129.59 g/mol
Frequently Bought Together