1,2,4-Trihydroxybenzene 95+%
Catalog # APOSOR5390-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:126.11 g/mol
Melting Pt:140 °C (subl.)
Storage Temperature:Refrigerator
CAS Number:533-73-3
Formula:C₆H₆O₃
MDL Number:MFCD00002198