
2,3-Dichlorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:82417-45-6
:
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00051844
:245.51 g/mol
Melting Pt:57…64 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together