
2,4-Dihydroxybenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64419-24-5
:
Formula:C₇H₅NO₂
MDL Number:MFCD02261931
:135.12 g/mol
Melting Pt:179 °C
Storage Temperature:Ambient
EINECS:264-886-2
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together