5-Nitro-1,2-benzisoxazole 97%
Catalog # APOSOR480767-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.1200 g/mol
Storage Temperature:Refrigerator
CAS Number:39835-28-4
Formula:C₇H₄N₂O₃
MDL Number:MFCD06659648