(Isopropylamino)(oxo)acetic acid
Catalog # APOSOR470739-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:131.13 g/mol
Storage Temperature:Ambient
CAS Number:29262-57-5
Formula:C₅H₉NO₃
MDL Number:MFCD06740186