4-[3-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)propyl]piperazine-1-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:857266-28-5
:
Formula:C₂₀H₂₇N₃O₄
MDL Number:MFCD09029311
:373.46 g/mol
Storage Temperature:Ambient
Frequently Bought Together