Order Entry
Spain
ContactUsLinkComponent
2,4-Dichlorothiophenol 98%
Irritant
2,4-Dichlorothiophenol 98%
Catalog # APOSOR3331-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dichlorothiophenol 98%
Catalog # APOSOR3331-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :179.07 g/mol
Boiling Pt:60…62 °C/1.5mmHg
Melting Pt:15 °C
Density:1.44 g/cm³ (25 °C)
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1122-41-4
Formula:C₆H₄Cl₂S
MDL Number:MFCD00041421