Order Entry
Spain
ContactUsLinkComponent
2-Amino-3,5-dichlorobenzonitrile 98%
Irritant
2-Amino-3,5-dichlorobenzonitrile 98%
Catalog # APOSOR3214-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-3,5-dichlorobenzonitrile 98%
Catalog # APOSOR3214-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.03 g/mol
Melting Pt:123…127 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:36764-94-0
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD00158947