
Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate
Catalog # APOSOR309161-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:257.33 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:194726-40-4
Formula:C₁₃H₂₃NO₄
MDL Number:MFCD07374389