

Sulphur trioxide pyridine complex
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26412-87-3
:
Formula:C₅H₅NO₃S
MDL Number:MFCD00012437
:159.16 g/mol
Melting Pt:155 °C
UN:3261
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together