Order Entry
Spain
ContactUsLinkComponent
2,5-Dichlorobenzonitrile 98%
Irritant
2,5-Dichlorobenzonitrile 98%
Catalog # APOSOR24097-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,5-Dichlorobenzonitrile 98%
Catalog # APOSOR24097-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :172.01 g/mol
Melting Pt:128…132 °C
Storage Temperature:Ambient
EINECS:244-518-7
UN:3439
ADR:6.1,III
 :GHS07
CAS Number:21663-61-6
Formula:C₇H₃Cl₂N
MDL Number:MFCD00019741